Order Entry
Austria
ContactUsLinkComponent
Potassium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Corrosive
Irritant
Potassium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0222.1000
Supplier:  Avantor
CAS Number:  
Corrosive
Irritant
Potassium hydroxide, BAKER ANALYZED® ACS, J.T.Baker®
Catalog # 0222.1000
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

Specification Test Results

  • Assay:
    min. 86.0 %
  • Calcium (Ca):
    max. 0.005 %
  • Chloride (Cl):
    max. 0.001 %
  • Magnesium (Mg):
    max. 0.002 %
  • Sodium (Na):
    max. 0.05 %
  • Heavy Metals (as Ag):
    max. 0.001 %
  • Insoluble Matter:
    max. 30 ppm
  • Potassium Carbonate (K2CO3):
    max. 0.5 %
  • Iron (Fe):
    max. 3
  • Mercury (Hg):
    max. 0.1
  • Nickel (Ni):
    max. 2
  • Nitrogen Compounds (as N):
    max. 3
  • Phosphate (PO4):
    max. 2
  • Sulfate (SO4):
    max. 5

Specification Test Results

About this item

 :
 :56.11 g/mol
Merck Index:12,07806
Boiling Pt:1320 °C (1013 hPa)
Melting Pt:360 °C
Density:2.04 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
 :GHS05
 :GHS07
CAS Number:1310-58-3
Formula:KOH
MDL Number:MFCD00003553