

Potassium hydroxide ≥99.98% (metals basis) 85% min (KOH basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1310-58-3
:
Formula:KOH
MDL Number:MFCD00003553
:56.11 g/mol
Merck Index:14,7640
Boiling Pt:1320…1324 °C
Melting Pt:360 °C
Density:2.044
Storage Temperature:Ambient
EINECS:215-181-3
UN:1813
ADR:8,II
REACH:01-2119487136-33
:GHS05
:GHS07
Related Information
Frequently Bought Together