Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Potassium hydroxide
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
:
Thermo Fisher Scientific
Potassium hydroxide ≥85% ACS (max. 2% K₂CO₃)
:
Thermo Fisher Scientific
:
1310-58-3
:
:
KOH
:
MFCD00003553
:
56.11 g/mol
:
14,7640
:
1320…1324 °C
:
360 °C
:
2.044
:
Ambient
:
215-181-3
:
1813
:
8,II
:
01-2119487136-33
:
GHS05
:
GHS07
Documentation
SDS
msds_ALFA013451
Videos
Thermo Scientific Acroseal 25 Years
Documentation
Frequently Bought Together
Test kits, ammonium, Spectroquant®, Supelco®
: Merck
pH indicator strips, MQuant®, Supelco®
: Merck
Supelco® Spectroquant® Test Kits, Sulphate
: Merck
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Test strips, sulphite, MQuant®, Supelco®
: Merck
Test kits, nitrogen (total) cell tests, Spectroquant®, Supelco®
: Merck
Test kits, surfactants, Spectroquant®, Supelco®
: Merck
M-FIX™ Spray fixative for gynecological and clinical cytology, Sigma-Aldrich®
: Merck
Visual colour kits, total hardness, MQuant®, Supelco®
: Merck
Test kits, phosphate, Spectroquant®, Supelco®
: Merck
VWR® Dosatest, pH indicator test strips, non bleeding
: VWR Chemicals
Test strips, peroxide, Dosatest
: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
: Merck
Test kits, chloride, Spectroquant®, Supelco®
: Merck
Test kits, nitrate, Spectroquant®, Supelco®
: Merck
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN
: Hach
Test kits, photometric, COD, LCK cuvette tests
: Hach