Sodium azide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sodium triazide
CAS Number:26628-22-8
:
Formula:NaN₃
MDL Number:MFCD00003536
Density:1.85 g/cm³ (20 °C)
Molecular Weight:65.01 g/mol
Melting Pt:275
ADR:6.1 II
Merck Index:13,08653
:M-Clarity™ quality level = MQ200
EINECS:247-852-1
UN:1687
:GHS06
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together