Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Austria
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Trisodium hexafluoroferrate
Trisodium hexafluoroferrate, Sigma-Aldrich®
Trisodium hexafluoroferrate, Sigma-Aldrich®
SIAL208965-25G
:
Merck
:
Trisodium hexafluoroferrate, Sigma-Aldrich®
SIAL208965-25G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
25 g
Seq:
3308
:
:
238.81 g/mol
:
244-124-5
:
20955-11-7
:
F₆FeNa₃
:
MFCD00042147