Lithium chloride 99%, Reagent Grade
Supplier: Honeywell Chemicals
Certificates
About this item
CAS Number:7447-41-8
:
Formula:LiCl
MDL Number:MFCD00011078
:42.39 g/mol
Merck Index:13,05550
Boiling Pt:1.360 °C (1.013 hPa)
Melting Pt:605 °C
Density:2.07 g/cm3 (20 °C)
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Assay:98.5 - 101.5 %
Specification Test Results
Frequently Bought Together