Order Entry
Austria
ContactUsLinkComponent
 
Potassium chloride ≥99.0%, Reagent Grade
Catalog # P4504-5KG
CAS Number:  
Potassium chloride ≥99.0%, Reagent Grade
Catalog # P4504-5KG
CAS Number:  

Specifications

  • Pack type:
    Drum with inliner
  • Pk:
    5 kg
  • Seq:
    4114

Specifications

Specification Test Results

  • Assay:
    99.0 - 101.0 %
  • Appearance of the solution:
    complying

Specification Test Results

About this item

 :
 :74.55 g/mol
Merck Index:14,07621
Boiling Pt:1.413 °C (1.013 hPa)
Melting Pt:772 °C
Density:2 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-211-8
CAS Number:7447-40-7
Formula:KCl
MDL Number:MFCD00011360