Cesium chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:7647-17-8
:
Formula:CsCl
MDL Number:MFCD00010955
:168.36 g/mol
Merck Index:13,02021
Boiling Pt:1303 °C (1013 hPa)
Melting Pt:646 °C
Density:3.7808 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:231-600-2
Frequently Bought Together