Order Entry
Austria
Orders LinkContactUsLinkComponent
Rubidium bromide, Sigma-Aldrich®

Rubidium bromide, Sigma-Aldrich®

Glass bottle for solids, 10 g
Catalog # SIAL336149-10G
Supplier:   Merck
Rubidium bromide, Sigma-Aldrich®
Catalog # SIAL336149-10G
Supplier:  Merck
CAS Number:  7789-39-1
Rubidium bromide, Sigma-Aldrich®

Glass bottle for solids, 10 g

CAS Number:  7789-39-1
Price
154,00 €
Log in for availability
1 * 10 g
Product Details & Documents
  • CAS Number:7789-39-1
  • Formula:BrRb
  • MDL Number:MFCD00011186
  • Molecular Weight:165.37 g/mol
  • Merck Index:12,08440
  • Boiling Pt:1340 °C (1013 hPa)
  • Melting Pt:693 °C
  • Density:3.35 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:232-162-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.