Order Entry
Austria
Orders LinkContactUsLinkComponent
Lithium bromide, Sigma-Aldrich®

Lithium bromide, Sigma-Aldrich®

Irritant
Supplier:   Merck
Lithium bromide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Lithium bromide, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
  • CAS Number:7550-35-8
  • Formula:LiBr
  • MDL Number:MFCD00011077
  • Molecular Weight:86.84 g/mol
  • Merck Index:13,05548
  • Boiling Pt:1265 °C (1013 hPa)
  • Melting Pt:550 °C
  • Density:3.46 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:231-439-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 78,60 €
    78,60 €
    1 * 5 g
       78,60 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 256,00 €
    256,00 €
    1 * 25 g
       256,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.