Order Entry
Austria
ContactUsLinkComponent
Sodium tetraphenylborate ≥99.5% analytical reagent
Toxic
Sodium tetraphenylborate ≥99.5% analytical reagent
Catalog # 28187.138
Supplier:  VWR Chemicals
CAS Number:  
undefined
Toxic
Sodium tetraphenylborate ≥99.5% analytical reagent
Catalog # 28187.138
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Assay:
    Min. 99.5 %
  • Loss on drying (100°C):
    Max. 0.5 %

Specification Test Results

About this item

 :
 :342.22 g/mol
Merck Index:13,08765
Melting Pt:> 400 °C
Density:1.15 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-605-5
UN:2811
ADR:6.1,III
 :GHS06
CAS Number:143-66-8
Formula:(C₆H₅)₄BNa
MDL Number:MFCD00011494