Order Entry
Austria
ContactUsLinkComponent
Sodium tetraphenylborate ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Toxic
Sodium tetraphenylborate ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates
Toxic
Sodium tetraphenylborate ACS, Reag. Ph. Eur. analytical reagent, Supelco®
Supplier:  Merck
Certificates

Specification Test Results

  • Solubility:
    (20 °C) soluble
  • Melting Point:
    300 °C
  • Molar Mass:
    342.22 g/mol
  • Bulk Density:
    500 kg/m3
  • pH:
    8 (50 g/l, H2O, 20 °C)
  • Hazard Class:
    6.1
  • Hazard Class Description:
    Toxic materials

Specification Test Results

Related Information

Certificates
Search by Lot Number
Frequently Bought Together