Order Entry
Austria
ContactUsLinkComponent
Lithium aluminum hydride, Sigma-Aldrich®
Flammable
Corrosive
Lithium aluminum hydride, Sigma-Aldrich®
Artikelnr. SIAL686034-10G
Supplier:  Merck
CAS Number:  
Flammable
Corrosive
Lithium aluminum hydride, Sigma-Aldrich®
Artikelnr. SIAL686034-10G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g
  • Seq:
    3236

Specifications

About this item

 :
 :37.95 g/mol
Merck Index:13,00348
Melting Pt:125 °C
Density:0.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-877-9
UN:1410
ADR:4.3,I
 :M-Clarity™ quality level = MQ100
 :GHS02
 :GHS05
CAS Number:16853-85-3
Formula:H₄AlLi
MDL Number:MFCD00011075