

Lithium aluminum hydride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:16853-85-3
:
Formula:H₄AlLi
MDL Number:MFCD00011075
:37.95 g/mol
Merck Index:13,00348
Melting Pt:125 °C
Density:0.91 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:240-877-9
ADR:4.3,I
:M-Clarity™ quality level = MQ200
UN:1410
:GHS02
:GHS05
Frequently Bought Together