5-(Hydroxymethyl)-2-furaldehyde 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:67-47-0
:
Formula:C₆H₆O₃
MDL Number:MFCD00003234
Merck Index:13,04857
Boiling Pt:114…116 °C (1 mmHg)
Melting Pt:30…34 °C
Density:1,24 g/cm³ (20 °C)
Flash Pt:79 °C
:126.11 g/mol
EINECS:200-654-9
:GHS07
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Colorless to white to yellow to orange to brown Liquid or crystalline powder and/or chunks or fused solid
- Infrared spectrum:Conforms
- Melting point:28 to 34 °C
- GC:≥97.5 %
Specification Test Results
Frequently Bought Together