Cyclohexylméthanol 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:100-49-2
:
Formula:C₇H₁₄O
MDL Number:MFCD00001510
:114.19 g/mol
Merck Index:13,02760
Boiling Pt:181 °C (760 mmHg)
Melting Pt:-43 °C
Density:0.91 g/cm³
Flash Pt:71 °C
Storage Temperature:Ambient
EINECS:202-857-8
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥98.5 %
- HPLC:≥96.0 %
- Specific optical rotation:-105° to -109° (20°C, 589 nm) (C=1, methanol)
Specification Test Results
Frequently Bought Together