Cyclobutanol, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2919-23-5
:
Formula:C₄H₈O
MDL Number:MFCD00001318
:72.11 g/mol
Boiling Pt:122 °C (1013 hPa)
Density:0.9226 g/cm³ (15 °C)
Storage Temperature:Ambient
EINECS:220-858-1
UN:1120
ADR:3,II
:M-Clarity™ quality level = MQ100
Frequently Bought Together