Order Entry
Austria
ContactUsLinkComponent
7-Amino-4-methylcoumarin, Sigma-Aldrich®
7-Amino-4-methylcoumarin, Sigma-Aldrich®
Catalog # 164545-10
CAS Number:  
7-Amino-4-methylcoumarin, Sigma-Aldrich®
Catalog # 164545-10
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :175.19 g/mol
Boiling Pt:352 °C (1013 hPa)
Melting Pt:226 °C
Density:1.377 g/cm³ (20 °C)
EINECS:247-454-8
CAS Number:26093-31-2
Formula:C₁₀H₉NO₂
MDL Number:MFCD00006868