Hydrochloric acid 1.128% (w/v) analytical reagent
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7647-01-0
:
Formula:HCl
MDL Number:MFCD00011324
:36.46 g/mol
Specification Test Results
- Assay (w/v):1.118 - 1.138 %
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_VWRC86132.290 spec_VWRC86132.290 spec_VWRC86132.290 spec_VWRC86132.360 spec_VWRC86132.360
Product specification
More
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together