Order Entry
Austria
ContactUsLinkComponent
2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)
2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)
Catalog # PROOCIL-PCB-197-C
CAS Number:  
2,2',3,3',4,4',6,6'-Octachlorobiphenyl (PCB No. 197)
Catalog # PROOCIL-PCB-197-C
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :429.77 g/mol
CAS Number:33091-17-7
Formula:C₁₂H₂Cl₈