2-Mesitylenesulphonyl chloride
Catalog # MOLE27848042-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
- Environmentally Preferable:
Specifications
About this item
Synonyms: 2,4,6-Trimethylbenzenesulphonyl chloride, Mesitylene-2-sulphonyl chloride
:
:218.7 g/mol
Boiling Pt:150 °C (20 mmHg)
Melting Pt:53…56 °C
Storage Temperature:Ambient
EINECS:212-257-8
UN:3261
ADR:8,II
:GHS05
CAS Number:773-64-8
Formula:C₉H₁₁ClO₂S
MDL Number:MFCD00007434