Order Entry
Austria
ContactUsLinkComponent
2,6-Di-tert-butyl-p-cresol analytical standard, Supelco®
2,6-Di-tert-butyl-p-cresol analytical standard, Supelco®
Catalog # 47168.
Supplier:  Merck
CAS Number:  
undefined
2,6-Di-tert-butyl-p-cresol analytical standard, Supelco®
Catalog # 47168.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • Seq:
    0001

Specifications

About this item

 :
 :220.35 g/mol
Merck Index:13,01547
Boiling Pt:265 °C (1013 hPa)
Melting Pt:69…70 °C
Density:1.05 g/cm³ (20 °C)
Flash Pt:127 °C
Storage Temperature:Ambient
EINECS:204-881-4
CAS Number:128-37-0
Formula:C₁₅H₂₄O
MDL Number:MFCD00011644