Order Entry
Austria
ContactUsLinkComponent
 
 
 
 
p-Cymene 99+%
 
undefined
 
 
 
p-Cymene 99+%
 

 

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4890 to 1.4910 (20°C, 589 nm)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤30 APHA
  • Specific gravity:
    (25°C) 0.853 to 0.860

 

 

Frequently Bought Together