2,3-Xylyl isothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
Synonyms: 2,3-Dimethylphenyl isothiocyanate
CAS Number:1539-20-4
:
Formula:C₉H₉NS
MDL Number:MFCD00041064
:163.24
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together