Order Entry
Austria
Orders LinkContactUsLinkComponent
3'-(Trifluoromethyl)acetophenone

3'-(Trifluoromethyl)acetophenone

Glass bottle for solids, 5 g
Catalog # MOLEM61706400
Irritant
Marketsource
Supplier:   Molekula
3'-(Trifluoromethyl)acetophenone
Catalog # MOLEM61706400
Supplier:  Molekula
Marketsource
CAS Number:  349-76-8
3'-(Trifluoromethyl)acetophenone

Glass bottle for solids, 5 g

CAS Number:  349-76-8
Price
Inquire For Price
Log in for availability
1 * 5 g
Product Details & Documents
Synonyms:  m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one
  • CAS Number:349-76-8
  • Formula:C₉H₇F₃O
  • MDL Number:MFCD00000391
  • Molecular Weight:188.15 g/mol
  • Boiling Pt:198 °C (1013 hPa)
  • Density:1,23 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:206-490-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g

Specifications