2'-(Trifluoromethyl)acetophenone, Sigma-Aldrich®
Catalog # SIAL233153-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
- Environmentally Preferable:
Specifications
About this item
Synonyms: o-(Trifluoromethyl)acetophenone, 1-[2-(Trifluoromethyl)phenyl]ethan-1-one, 1-(2-(Trifluoromethyl)phenyl)ethanone
:
:188.15 g/mol
Boiling Pt:163 °C (1013 hPa)
Melting Pt:15…17 °C
Density:1.25 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:241-434-2
CAS Number:17408-14-9
Formula:C₉H₇F₃O
MDL Number:MFCD00000378