Thymol blue ACS
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:76-61-9
:
Formula:C₂₇H₃₀O₅S
MDL Number:MFCD00005869
:466.6 g/mol
Merck Index:14,9400
Melting Pt:223 °C (dec.)
Storage Temperature:Ambient
EINECS:200-973-3
Related Information
Frequently Bought Together