Bromothymol blue ACS, Reag. Ph. Eur.
Supplier: Merck
Certificates
About this item
CAS Number:76-59-5
:
Formula:C₂₇H₂₈Br₂O₅S
MDL Number:MFCD00005872
Merck Index:13,01430
Melting Pt:204…202 °C
:M-Clarity™ quality level = MQ200
:624.39 g/mol
Storage Temperature:Ambient
EINECS:200-971-2
Related Information
Certificates
Search by Lot NumberMERC1.03026.0025_TDS_EN ProductPage_103026
Product specification
Merck Brighter titration Titripur®
Product Literature
msds_103026
SDS
Frequently Bought Together