ContactUsLinkComponent
Bromothymol blue sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Bromothymol blue sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Certificates
Bromothymol blue sultone form ACS, Reag. Ph. Eur. indicator, Fluka™
Certificates
Specification Test Results
  • Suitability as indicator:
    complying
  • Test of sensitivity:
    complying
  • Clearness of the solution:
    complying
  • Change Range (pH6,0-7,4):
    complying
  • Curve course (UV):
    complying