Order Entry
Austria
ContactUsLinkComponent
Salicyl alcohol for synthesis, Sigma-Aldrich®
Salicyl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Salicyl alcohol for synthesis, Sigma-Aldrich®
Supplier:  Merck

Specification Test Results

  • Solubility:
    63 g/l (20 °C)
  • Melting Point:
    83 - 86 °C
  • Molar Mass:
    124.14 g/mol
  • Synonyms:
    Saligenine, Salicyl alcohol
  • Hill Formula:
    C7H8O2
  • Chemical Formula:
    2-(OH)C6H4CH2OH

Specification Test Results

Related Information

Frequently Bought Together