Order Entry
Austria
Orders LinkContactUsLinkComponent
m-Cresol ≥99%

m-Cresol ≥99%

Glass bottle for solids, 250 g
Catalog # A10733.30
Toxic
Corrosive
m-Cresol ≥99%
Catalog # A10733.30
CAS Number:  108-39-4
m-Cresol ≥99%
m-Cresol ≥99%

Glass bottle for solids, 250 g

CAS Number:  108-39-4
Price
36,90 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 108-39-4 Molecular Formula: C7H8O Formula Weight: 108.14 Flash Point: 86°(186°F) Refractive Index: 1.5400 Odour: Phenol-like pH: 5 Melting Point: 11-12° Boiling Point: 202-204° Solubility: Soluble in water, alcohol, ether, chloroform Beilstein Reference: 506719 Merck Reference: 14,2579 UN No.: 2076 MDL No.: MFCD00002302

Synonyms:  1-Hydroxy-3-methylbenzene, 3-Methylphenol, 3-Hydroxytoluol, 3-Hydroxytoluene
  • CAS Number:108-39-4
  • Formula:CH₃C₆H₄OH
  • MDL Number:MFCD00002302
  • Molecular Weight:108.14 g/mol
  • Merck Index:14,2579
  • Boiling Pt:202…204 °C
  • Melting Pt:11…12 °C
  • Density:1.034
  • Flash Pt:86 °C (186 °F)
  • Storage Temperature:Ambient
  • EINECS:203-577-9
  • UN:2076
  • ADR:6.1,II
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications