
2-Phenylethanol for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:60-12-8
:
Formula:C₆H₅CH₂CH₂OH
MDL Number:MFCD00002886
:122.17 g/mol
Merck Index:12,07370
Boiling Pt:432 °C (1013 hPa)
Melting Pt:103…105 °C
Density:1.02 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:200-456-2
UN:2810
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:102 °C
:GHS07
Specification Test Results
- Solubility:20 g/l (20 °C)
- Melting Point:-27 °C
- Molar Mass:122.16 g/mol
- Boiling Point:219 °C (1013 hPa)
- Vapor Pressure:0.08 hPa (20 °C)
- Flash Point:101.5 °C
- Refractive Index:1.5325 (20 °C, 589 nm)
- Explosion Limit:1.4 - 11.9 %(V)
- Density:1.02 g/cm3 (20 °C)
- pH:6 - 7 (20 g/l, H2O, 20 °C)
- Ignition Point:410 °C
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Specification Test Results
Related Information
Frequently Bought Together