Order Entry
Austria
ContactUsLinkComponent
 
Toxic
Health
Environment
3-Nitro-o-toluidine
Catalog # EHERC10207700
Supplier:  EHRENSTORFER
CAS Number:  
Toxic
Health
Environment
3-Nitro-o-toluidine
Catalog # EHERC10207700
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
Boiling Pt:305 °C (1013 hPa)
Melting Pt:87…89 °C
Density:0.33 g/cm³ (20 °C)
ADR:6.1,III
 :152.15 g/mol
EINECS:210-059-6
UN:2660
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:603-83-8
Formula:C₇H₈N₂O₂
MDL Number:MFCD00007731