p-Toluidine 99%, crystalline molten mass
: 4-Aminotoluene, 4-Methylaniline, 4-Toluidine
:106-49-0
:
:C₇H₉N
:MFCD00007906
:200 °C (760 mmHg)
:107.16 g/mol
:41…46 °C
:87 °C
:6.1 II
:12,09674
:203-403-1
:3451
:GHS06
:GHS08
:GHS09
:Ambient
- Environmentally Preferable:
- Appearance:Colorless to red Liquid after melting or solid (May discolor to brown on storage)
- Infrared spectrum:Conforms
- Melting point:41 to 46 °C
- GC:≥98.5 %
Frequently Bought Together