o-Toluidine, Sigma-Aldrich®
Catalog # SIALT1199-500ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
About this item
:
:107.16 g/mol
Merck Index:12,09674
Boiling Pt:200 °C (1013 hPa)
Melting Pt:< –15 °C
Density:0.998 g/cm³ (20 °C)
Flash Pt:85 °C
Storage Temperature:Ambient
EINECS:202-429-0
UN:1708
ADR:6.1,II
:M-Clarity™ quality level = MQ200
:GHS06
:GHS08
:GHS09
CAS Number:95-53-4
Formula:C₇H₉N
MDL Number:MFCD00007730