Order Entry
Austria
ContactUsLinkComponent
o-Toluidine
o-Toluidine
Catalog # EHERC17594800
Supplier:  EHRENSTORFER
CAS Number:  
o-Toluidine
Catalog # EHERC17594800
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • Seq:
    2070

Specifications

About this item

 :
 :107.16 g/mol
Merck Index:12,09674
Boiling Pt:200 °C (1013 hPa)
Melting Pt:< –15 °C
Density:0.998 g/cm³ (20 °C)
Flash Pt:85 °C
EINECS:202-429-0
UN:1708
ADR:6.1,II
 :GHS06
 :GHS08
 :GHS09
Storage Temperature:Ambient
CAS Number:95-53-4
Formula:C₇H₉N
MDL Number:MFCD00007730