Order Entry
Austria
ContactUsLinkComponent
o-Toluidine 99%
o-Toluidine 99%
Certificates
undefined
o-Toluidine 99%

Specification Test Results

  • Appearance:
    Clear yellow to orange to light brown Liquid (May darken on storage)
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5710 to 1.5730 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specification Test Results

Related Information

Frequently Bought Together