Order Entry
Austria
Orders LinkContactUsLinkComponent
2',5'-Dihydroxyacetophenone

2',5'-Dihydroxyacetophenone

Glass bottle for solids, 25 g
Catalog # MOLEM45596894
Marketsource
Supplier:   Molekula
2',5'-Dihydroxyacetophenone
Catalog # MOLEM45596894
Supplier:  Molekula
Marketsource
CAS Number:  490-78-8
2',5'-Dihydroxyacetophenone

Glass bottle for solids, 25 g

CAS Number:  490-78-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone
  • CAS Number:490-78-8
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00002343
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:199…202 °C
  • EINECS:207-716-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Cat. No.:
    MOLEM45596894

Specifications