Order Entry
Austria
Orders LinkContactUsLinkComponent
2',5'-Dihydroxyacetophenone 97%

2',5'-Dihydroxyacetophenone 97%

Irritant
2',5'-Dihydroxyacetophenone 97%
Certificates
2',5'-Dihydroxyacetophenone 97%

2 Options Available Below

Product Details & Documents
Synonyms:  1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone
  • CAS Number:490-78-8
  • Formula:C₈H₈O₃
  • MDL Number:MFCD00002343
  • Molecular Weight:152.15 g/mol
  • Boiling Pt:296 °C (1013 hPa)
  • Melting Pt:203…208 °C
  • EINECS:207-716-4

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Yellow to yellow-green Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    203 to 208 °C (Dec)
  • HPLC:
    ≥96.0 %
  • Clarity of solution:
    passes test
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,10 €
    29,10 €
    1 * 5 g
       29,10 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 84,20 €
    84,20 €
    1 * 25 g
       84,20 €