2-Bromo-4-isopropylaniline
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:51605-97-1
:
Formula:C₉H₁₂BrN
MDL Number:MFCD00051585
:214.1 g/mol
Boiling Pt:139…141 °C (20 mmHg)
Density:1.35 g/cm³ (20 °C)
UN:2810
ADR:6.1,III
Frequently Bought Together