ω-Bromo-4-fluoroacetophenone
Glass bottle for solids, 25 g
Catalog # MOLE17125731-25G
Marketsource
Supplier: Molekula
ω-Bromo-4-fluoroacetophenone
Catalog # MOLE17125731-25G
Supplier: Molekula
Marketsource
CAS Number: 403-29-2
Jump to:
Product Details & Documents
Synonyms: Bromomethyl p-fluorophenyl ketone, α-Bromo-4-fluoroacetophenone, 2-Bromo-1-(4-fluorophenyl)ethan-1-one, 2-Bromo-4'-fluoroacetophenone, 4-Fluorophenacyl bromide, 2-Bromo-1-(4-fluorophenyl)ethanone
- CAS Number:403-29-2
- Formula:C₈H₆BrFO
- MDL Number:MFCD00040830
- Molecular Weight:217.04 g/mol
- Boiling Pt:82 °C (0,4 mmHg)
- Boiling Pt:82 °C (0.4 mmHg)
- Melting Pt:49 °C
- EINECS:206-955-1
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



