(4-Bromophenyl)acetonitrile, Sigma-Aldrich®
Catalog # SIAL124664-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:196.05 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:47…49 °C
ADR:6.1,I
:M-Clarity™ quality level = MQ200
EINECS:240-602-2
UN:3449
CAS Number:16532-79-9
Formula:C₈H₆BrN
MDL Number:MFCD00001916