(3-Bromophenyl)acetonitrile
Catalog # MOLEM46799611
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:196.05 g/mol
Boiling Pt:286 °C (1013 hPa)
Melting Pt:27…28 °C
EINECS:250-867-6
UN:3449
ADR:6.1,I
CAS Number:31938-07-5
Formula:C₈H₆BrN
MDL Number:MFCD00001906