Order Entry
Austria
ContactUsLinkComponent
 
Sulphanilamide Reag. Ph. Eur. analytical reagent, Supelco®
  1.11799.0100
 :  Merck
 :  
undefined
Sulphanilamide Reag. Ph. Eur. analytical reagent, Supelco®
  1.11799.0100
 :  Merck
 :  

 

  • Pack type:
    Plastic bottle
  • Pk:
    100 g
  • Seq:
    3401

 

 

  • Solubility:
    7.5 g/l (25 °C)
  • Melting Point:
    163 - 167 °C
  • Molar Mass:
    172.2 g/mol
  • Bulk Density:
    500 kg/m3
  • Density:
    1.50 g/cm3
  • pH:
    5.8 - 6.1 (5 g/l, H2O, 20 °C)

 

 

 :
 :172.21 g/mol
 :13,09010
 :401 °C (1013 hPa)
 :163…167 °C
 :1.46 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :Ambient
 :200-563-4
 :63-74-1
 :H₂NC₆H₄SO₂NH₂
 :MFCD00007939