p-Anisidine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: 4-Anisidine, 4-Aminoanisole, 4-Methoxyaniline
CAS Number:104-94-9
:
Formula:C₇H₉NO
MDL Number:MFCD00007864
:123.15 g/mol
Merck Index:13,00670
Boiling Pt:243 °C (1013 hPa)
Melting Pt:55…58 °C
Density:1.2 g/cm³ (20 °C)
EINECS:203-254-2
UN:2431
ADR:6.1,III
:M-Clarity™ quality level = MQ200
Flash Pt:122 °C
:GHS06
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together