
N,N-Dibutylaniline 97%, Sigma-Aldrich®
:613-29-6
:
:(C₆H₅)N(CH₂CH₂CH₂CH₃)₂
:MFCD00035799
:205.34 g/mol
:265…275 °C (1013 hPa)
:–32 °C
:0.90368 g/cm³ (20 °C)
:Room Temperature
:210-335-6
:2810
:6.1,III
:GHS07
- Appearance (Colour):Colourless to Yellow to Green
- Appearance (Form):Liquid
- Infrared spectrum:Conforms to Structure
- Purity (GC):≥96.5%
Frequently Bought Together