Order Entry
Austria
ContactUsLinkComponent
p,α,α,α-Tetrafluoroanisole for synthesis, Sigma-Aldrich®
Flammable
p,α,α,α-Tetrafluoroanisole for synthesis, Sigma-Aldrich®
Catalog # 8.41175.0001
Supplier:  Merck
CAS Number:  
Flammable
p,α,α,α-Tetrafluoroanisole for synthesis, Sigma-Aldrich®
Catalog # 8.41175.0001
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 ml
  • Seq:
    2070

Specifications

Specification Test Results

  • Molar Mass:
    180.1 g/mol
  • Boiling Point:
    104 - 105 °C (1013 hPa)
  • Flash Point:
    15 °C
  • Density:
    1.323 g/cm3

Specification Test Results

About this item

 :
 :180.1 g/mol
Boiling Pt:104…105 °C (1013 hPa)
Density:1.323 g/cm³ (20 °C)
Storage Temperature:Ambient
ADR:3,II
 :M-Clarity™ quality level = MQ200
EINECS:206-523-2
UN:1993
 :GHS02
CAS Number:352-67-0
Formula:C₇H₄F₄O
MDL Number:MFCD00040835