Order Entry
Austria
ContactUsLinkComponent
4-(Trifluoromethoxy)aniline
4-(Trifluoromethoxy)aniline
Catalog # EHERC17844810
Supplier:  EHRENSTORFER
CAS Number:  
4-(Trifluoromethoxy)aniline
Catalog # EHERC17844810
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :177.13 g/mol
Boiling Pt:73 °C (1013 hPa)
Density:1.31 g/cm³ (20 °C)
EINECS:207-317-5
UN:2810
ADR:6.1,II
CAS Number:461-82-5
Formula:C₇H₆F₃NO
MDL Number:MFCD00041314