Storage boxes, LIGHT BOX
Supplier: MULTIROIR
Jump to:
About this item
Made of polypropylene.
- Totally transparent
- Slip lid with secure clip locking system
- Shock resistant
These storage boxes are perfectly suited to the storage and organisation of differently sized products. The contents can be easily identified through the transparent material.
9 Options Available Below
Product Details & Documents
Made of polypropylene.
- Totally transparent
- Slip lid with secure clip locking system
- Shock resistant
These storage boxes are perfectly suited to the storage and organisation of differently sized products. The contents can be easily identified through the transparent material.
Product Family Options
Product Information
- CapacityW×D×HAvailabilityPrice
- 1,7 LExternal: 205×170×75 mm
Top (internal): 180W×150D mm
Bottom (internal): 170W×140D mmLog in for availability1 * 5 ST27,20 € - 2,3 LExternal: 205×170×100 mm
Top (internal): 180W×150D mm
Bottom (internal): 165W×135D mmLog in for availability1 * 5 ST29,80 € - 3,5 LExternal: 205×170×145 mm
Top (internal): 180W×150D mm
Bottom (internal): 155W×125D mmLog in for availability1 * 5 ST33,40 € - 9,0 LExternal: 328×250×190 mm
Top (internal): 270W×205D mm
Bottom (internal): 240W×180D mmLog in for availability1 * 5 ST72,70 € - 15,0 LExternal: 380×285×217 mm
Top (internal): 320W×240D mm
Bottom (internal): 290W×210D mmLog in for availability1 * 5 ST89,30 € - 20,0 LExternal: 425×330×213 mm
Top (internal): 370W×285D mm
Bottom (internal): 335W×225D mmLog in for availability1 * 5 ST111,00 € - 30,0 LExternal: 595×395×170 mm
Top (internal): 530W×350D mm
Bottom (internal): 500W×320D mmLog in for availability1 * 5 ST167,00 € - 52,0 LExternal: 595×395×310 mm
Top (internal): 530W×350D mm
Bottom (internal): 490W×305D mmLog in for availability1 * 5 ST208,00 € - 134,0 LExternal: 780×570×400 mm
Top (internal): 705W×530D mm
Bottom (internal): 650W×470D mmLog in for availability1 * 5 ST601,00 €