
Phthalic acid, Sigma-Aldrich®
Catalog # SIALP39303-100G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:166.13 g/mol
Merck Index:12,07527
Boiling Pt:315…317 °C (1013 hPa)
Melting Pt:79 °C
Density:1.576 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:201-873-2
:GHS07
CAS Number:88-99-3
Formula:C₈H₆O₄
MDL Number:MFCD00002467